accessibility menu, dialog, popup

UserName

OOPOHOOHOHOHOHHONaNa

CAS RN 83833-15-2

IUPAC Name: {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}phosphonic acid disodium
Synonyms: glucose-1-phosphate disodium
Molecular Weight (g/mol): 306.11
Molecular Formula: C6H13Na2O9P
InChi Key: AQMGMJDBUKKFOG-UHFFFAOYNA-N
SMILES: [Na].[Na].OCC1OC(OP(O)(O)=O)C(O)C(O)C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results