missing translation for 'onlineSavingsMsg'
Learn More

beta-D-Glucopyranose 1-Phosphate Disodium Salt 98.0+%, TCI America™

Catalog No. G033920MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G033920MG 20 mg
G0339100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G033920MG Supplier TCI America Supplier No. G033920MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 83833-15-2
Molecular Formula C6H13Na2O9P
Molecular Weight (g/mol) 306.114
InChI Key AQMGMJDBUKKFOG-UZUGEDCSSA-N
PubChem CID 131885723
IUPAC Name sodium;[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
SMILES C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.[Na].[Na]

Specifications

Color White
Quantity 20 mg
Formula Weight 304.10
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material beta-D-Glucopyranose 1-Phosphate Disodium Salt
EINECSNumber (8)-0519
TSCA No
Recommended Storage Freezer