missing translation for 'onlineSavingsMsg'
Learn More

beta-D-Allose, 97+%

Catalog No. AAA16810MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16810MC 100 mg
AAA16810ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA16810MC Supplier Thermo Scientific Chemicals Supplier No. A16810MC

CAS: 2595-97-3 | C6H12O6 | 180.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2595-97-3
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00063268
InChI Key GZCGUPFRVQAUEE-OBOOZECYSA-N
PubChem CID 57449163
IUPAC Name (2R,3R,4R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 143°C to 146°C
Quantity 100 mg
Beilstein 1724623
Sensitivity Hygroscopic
Merck Index 14,280
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material beta-D-Allose
EINECSNumber 219-994-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only