missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Beta-casein Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149776
Description
Beta-casein Polyclonal Antibody for Western Blot
This gene is a member of the beta casein family. There are two types of casein protein, beta (encoded by this gene) and kappa, both of which are secreted in human milk. Beta casein is the principal protein in human milk and the primary source of essential amino acids for a suckling infant. Beta and kappa casein proteins acting together form spherical micelles which bind within them important dietary minerals, such as calcium and phosphorous. Alternative splicing results in multiple transcript variants.Chemical Identifiers
| Beta-casein | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P02666 | |
| 281099 | |
| Recombinant Casein Beta (CSN2) (Arg16-Val224). | |
| Antibody |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Bovine | |
| CSN2 | |
| beta casein; beta-caesin; beta-casein; beta-casein A3; beta-casein precursor; CASB; casein beta; CSN2; kappa-casein; Unknown (protein for MGC:133765); unnamed protein product | |
| CSN2 | |
| Primary | |
| Protein A |
Specifications
| Beta-casein | |
| Polyclonal | |
| Unconjugated | |
| CSN2 | |
| beta casein; beta-caesin; beta-casein; beta-casein A3; beta-casein precursor; CASB; casein beta; CSN2; kappa-casein; Unknown (protein for MGC:133765); unnamed protein product | |
| Rabbit | |
| Protein A | |
| RUO | |
| 281099 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P02666 | |
| CSN2 | |
| Recombinant Casein Beta (CSN2) (Arg16-Val224). | |
| 200 μL | |
| Primary | |
| Bovine | |
| Antibody |