missing translation for 'onlineSavingsMsg'
Learn More

beta-Bromo-4-fluorophenetole 98.0+%, TCI America™

Catalog No. p-7123501
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43765G 5 g
B437625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43765G Supplier TCI America Supplier No. B43765G
Only null left

Chemical Identifiers

CAS 332-48-9
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.05
MDL Number MFCD00044739
InChI Key JXSPKRUNMHMICQ-UHFFFAOYSA-N
Synonym 1-2-bromoethoxy-4-fluorobenzene, 4-fluorophenoxy-ethylbromide, 2-bromoethyl 4-fluorophenyl ether, 4-fluorophenoxyethylbromide, 4-2-bromoethoxy fluorobenzene, 4-fluorophenoxyethyl bromide, beta-bromo-4-fluorophenetole, 2-4-fluorophenoxy ethyl bromide, benzene, 1-2-bromoethoxy-4-fluoro, p-fluorophenoxy ethylbromide
PubChem CID 2064171
IUPAC Name 1-(2-bromoethoxy)-4-fluorobenzene
SMILES FC1=CC=C(OCCBr)C=C1

Specifications

Color Yellow
Boiling Point 66°C
Quantity 5 g
Formula Weight 219.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material beta-Bromo-4-fluorophenetole