missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ beta Alanine Polyclonal Antibody

Catalog No. PIPA5143449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143449 Supplier Invitrogen™ Supplier No. PA5143449
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The design and synthesis of beta-amino acids and of other biologically active amino acid derivatives is a challenge in current drug design The only commonly naturally occurring beta-amino acid is beta-alanine Beta Alanine is used as nutrition supplements in food production Increase muscular strength & power output, Increases Muscle Mass, increase Anaerobic Endurance.

Specifications

Antigen beta Alanine
Applications Dot Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Gene Alias 3-Aminopropanoic acid
Host Species Rabbit
Immunogen Beta Alanine.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less