missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Alanine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™
Supplier: TCI America A142325G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 27019-47-2 | |
| 180.23 | |
| CANCPUBPPUIWPX-UHFFFAOYSA-O | |
| 13502100 | |
| [NH3+]CCC(=O)OCC1=CC=CC=C1 |
| C10H14NO2 | |
| MFCD00039061 | |
| beta-alanine benzyl ester p-toluenesulfonate, beta-alanine benzyl ester p-toluenesulfonate salt, h-beta-ala-obzl.tosoh, h-beta-ala-obzl tos, benzyl 3-aminopropanoate 4-methylbenzenesulfonate, betaala-obzl.tosoh, pubchem12070, acmc-209gum, h-, a-ala-obzl.tosoh, h-ala-obzl p-tosylate | |
| 3-(benzyloxy)-3-oxopropan-1-aminium |
Specifications
| beta-Alanine Benzyl Ester p-Toluenesulfonate | |
| Yellow | |
| MFCD00039061 | |
| CANCPUBPPUIWPX-UHFFFAOYSA-O | |
| 3-(benzyloxy)-3-oxopropan-1-aminium | |
| 180.23 | |
| 351.42 | |
| Crystalline Powder |
| 27019-47-2 | |
| C10H14NO2 | |
| beta-alanine benzyl ester p-toluenesulfonate, beta-alanine benzyl ester p-toluenesulfonate salt, h-beta-ala-obzl.tosoh, h-beta-ala-obzl tos, benzyl 3-aminopropanoate 4-methylbenzenesulfonate, betaala-obzl.tosoh, pubchem12070, acmc-209gum, h-, a-ala-obzl.tosoh, h-ala-obzl p-tosylate | |
| [NH3+]CCC(=O)OCC1=CC=CC=C1 | |
| 25 g | |
| 13502100 | |
| ≥98.0% (HPLC,T) |
Safety and Handling
TSCA : No