missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. p-7117426
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14235G 5 g
A142325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14235G Supplier TCI America Supplier No. A14235G

Chemical Identifiers

CAS 27019-47-2
Molecular Formula C10H14NO2
Molecular Weight (g/mol) 180.23
MDL Number MFCD00039061
InChI Key CANCPUBPPUIWPX-UHFFFAOYSA-O
Synonym beta-alanine benzyl ester p-toluenesulfonate, beta-alanine benzyl ester p-toluenesulfonate salt, h-beta-ala-obzl.tosoh, h-beta-ala-obzl tos, benzyl 3-aminopropanoate 4-methylbenzenesulfonate, betaala-obzl.tosoh, pubchem12070, acmc-209gum, h-, a-ala-obzl.tosoh, h-ala-obzl p-tosylate
PubChem CID 13502100
IUPAC Name 3-(benzyloxy)-3-oxopropan-1-aminium
SMILES [NH3+]CCC(=O)OCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 351.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material beta-Alanine Benzyl Ester p-Toluenesulfonate