missing translation for 'onlineSavingsMsg'
Learn More

PeproTech beta-3 Defensin Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P24150UG
Encompass
Change view
Click to view available options
Quantity:
100 μg
50 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P24150UG 50 μg
500P241100 100 μg
500P2411MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 500P24150UG Supplier PeproTech Supplier No. 500P24150UG
Only null left

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: GIINTLQKYY CRVRGGRCAV LSCLPKEEQI GKCSTRGRKC CRRKK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BD-3. Anti-Human BD-3-specific antibody was purified by affinity chromatography employing an immobilized Human BD-3 matrix. Sandwich ELISA: To detect Human BD-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human BD-3 (500-P241Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BD-3. Western Blot: To detect Human BD-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BD-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].

Specifications

Antigen beta-3 Defensin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DEFB103A
Gene Accession No. P81534
Gene Alias BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103A; defensin, beta 103; defensin, beta 103A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3
Gene Symbols DEFB103A
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human BD-3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 414325
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less