missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ beta-2 Adaptin Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670134
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01670134 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01670134 Supplier Invitrogen™ Supplier No. PA11066
Season of Savings
View all promotions

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-1066 detects beta Adaptin 2 from human, mouse, rat, and bovine samples. PA1-1066 has been successfully used in Western blot procedures. The PA1-1066 immunogen is a synthetic peptide corresponding to residues I(588) H R K H L P I H H G S T D A G D S P(606) of human beta Adaptin 2. PA1-1066 can be used with blocking peptide PEP-293.

Clathrin-mediated endocytosis is the pathway by which many receptors for nutrients and hormones are internalized to be recycled or down-regulated. During formation of clathrin coated membranes, clathrin co-assembles with heterotetrameric molecules known as assembly polypeptides (APs) or adaptors which form a layer of protein coat between the clathrin lattice and the membrane. There are two characterized adaptors AP1 and AP2. AP1 is associated with clathrin coated vesicles at the trans-Golgi network and AP2 is associated with the endocytic clathrin coated vesicles at the plasma membrane and has been shown to specifically interact with Shc and EGF receptor. AP2 is composed of four subunits, two separate ~100 kDa gene products with similar domain structures (alpha and beta adaptin) and a ~50 and ~17 kDa subunit. There are two alpha-adaptin genes, alpha A and alpha C which have a tissue specific pattern of expression.

Specifications

Antigen beta-2 Adaptin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene AP2B1
Gene Accession No. P62944, P63010, Q08DS7, Q9DBG3
Gene Alias 1300012O03Rik; adapter-related protein complex 1 subunit beta-1; adapter-related protein complex 2 beta subunit; adapter-related protein complex 2 subunit beta; adaptin, beta 2 (beta); adaptor protein complex AP-1 subunit beta-1; Adaptor protein complex AP-2 subunit beta; adaptor related protein complex 2 beta 1 subunit; adaptor related protein complex 2 subunit beta 1; adaptor related protein complex 2, beta 1 subunit; adaptor-related protein complex 1 subunit beta-1; Adaptor-related protein complex 2 subunit beta; adaptor-related protein complex 2, beta 1 subunit; ADTB2; AI788979; AP-1 complex subunit beta-1; AP105B; AP-2 complex subunit beta; Ap2b1; AP2-BETA; beta adaptin; beta-1-adaptin; beta1-adaptin; Beta-2-adaptin; beta2-adaptin; beta-adaptin; Beta-adaptin 1; CLAPB1; Clathrin assembly protein complex 1 beta large chain; clathrin assembly protein complex 2 beta large chain; clathrin-associated / assembly / adaptor protein, large, beta 1; clathrin-associated/assembly/adaptor; clathrin-associated/assembly/adaptor protein, large, beta 1; golgi adaptor HA1/AP1 adaptin beta subunit; Plasma membrane adaptor HA2/AP2 adaptin beta subunit; testicular tissue protein Li 22
Gene Symbols AP2B1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues I(588) H R K H L P I H H G S T D A G D S P(606) of human beta Adaptin 2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140670, 163, 282183, 71770
Target Species Human, Mouse, Rat, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less