Learn More
Bestatin
Description
Bestatin can indirectly act on monocytes by inhibiting Aminopeptidase B, which in turn inhibits the catabolism of tuftsin. Bestatin has also been shown to inhibit Leukotriene A4 hydrolase. It can directly stimulate lymphocytes through its fixation on the cell surface. The agent has been used in multiple leukemia studies. Bestatin is an aminopeptidase inhibitor, inhibits enkephalin metabolism and leukotriene A4 hydrolase. Inhibits tumor cell proliferation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bestatin can indirectly act on monocytes by inhibiting Aminopeptidase B, which in turn inhibits the catabolism of tuftsin. Bestatin has also been shown to inhibit Leukotriene A4 hydrolase. It can directly stimulate lymphocytes through its fixation on the cell surface. The agent has been used in multiple leukemia studies. Bestatin is an aminopeptidase inhibitor, inhibits enkephalin metabolism and leukotriene A4 hydrolase. Inhibits tumor cell proliferation.
Solubility
Soluble in water (<1 25), DMSO (2 mg/ml), methanol (5 mg/ml), 1eq. NaOH (50 mM), ethanol (<1mg/mL at 25°C), acetic acid, aq. HCl, and alkaline solution. Insoluble in ethyl acetate, benzene, hexane, and chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 233°C to 236°C |
| Color | White |
| Quantity | 100 mg |
| Merck Index | 14,9842 |
| Solubility Information | Soluble in water (<1 25),DMSO (2mg/ml),methanol (5mg/ml),1eq. NaOH (50 mM),ethanol (<1mg/ml at 25°C),acetic acid,aq. HCl,and alkaline solution. Insoluble in ethyl acetate,benzene,hexane,and chloroform. |
| Formula Weight | 308.37 |
| Physical Form | Powder |
| Chemical Name or Material | Bestatin |
Safety and Handling
| Hazard Category | H315-H319-H335-H373 |
| Precautionary Statement | P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c |
| EINECSNumber | 261-529-2 |
| RTECSNumber | OH2915000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only