missing translation for 'onlineSavingsMsg'
Learn More

Bestatin

Catalog No. AAJ61106MC
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61106MC 100 mg
AAJ61106MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61106MC Supplier Thermo Scientific Chemicals Supplier No. J61106MC

CAS: 58970-76-6 | C16H24N2O4 | 308.38 g/mol

Bestatin can indirectly act on monocytes by inhibiting Aminopeptidase B, which in turn inhibits the catabolism of tuftsin. Bestatin has also been shown to inhibit Leukotriene A4 hydrolase. It can directly stimulate lymphocytes through its fixation on the cell surface. The agent has been used in multiple leukemia studies. Bestatin is an aminopeptidase inhibitor, inhibits enkephalin metabolism and leukotriene A4 hydrolase. Inhibits tumor cell proliferation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bestatin can indirectly act on monocytes by inhibiting Aminopeptidase B, which in turn inhibits the catabolism of tuftsin. Bestatin has also been shown to inhibit Leukotriene A4 hydrolase. It can directly stimulate lymphocytes through its fixation on the cell surface. The agent has been used in multiple leukemia studies. Bestatin is an aminopeptidase inhibitor, inhibits enkephalin metabolism and leukotriene A4 hydrolase. Inhibits tumor cell proliferation.

Solubility
Soluble in water (<1 25), DMSO (2 mg/ml), methanol (5 mg/ml), 1eq. NaOH (50 mM), ethanol (<1mg/mL at 25°C), acetic acid, aq. HCl, and alkaline solution. Insoluble in ethyl acetate, benzene, hexane, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 58970-76-6
Molecular Formula C16H24N2O4
Molecular Weight (g/mol) 308.38
MDL Number MFCD00083262
InChI Key VGGGPCQERPFHOB-MCIONIFRSA-N
Synonym bestatin, ubenimex, ubenimexum latin, ubenimex inn:jan, bestatin;ubenimex, s-2-2s,3r-3-amino-2-hydroxy-4-phenylbutanamido-4-methylpentanoic acid, ubenimex bestatin, chembl29292, 2s,3r-3-amino-2-hydroxy-4-phenylbutanoyl-l-leucine, 3-r-amino-2-s-hydroxy-4-phenylbutanoyl-s-leucine
PubChem CID 72172
IUPAC Name (2S)-2-[[(2S,3R)-3-amino-2-hydroxy-4-phenylbutanoyl]amino]-4-methylpentanoic acid
SMILES CC(C)C[C@@H](NC(=O)[C@@H](O)[C@H](N)CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 233°C to 236°C
Color White
Quantity 100 mg
Merck Index 14,9842
Solubility Information Soluble in water (<1 25),DMSO (2mg/ml),methanol (5mg/ml),1eq. NaOH (50 mM),ethanol (<1mg/ml at 25°C),acetic acid,aq. HCl,and alkaline solution. Insoluble in ethyl acetate,benzene,hexane,and chloroform.
Formula Weight 308.37
Physical Form Powder
Chemical Name or Material Bestatin
Hazard Category H315-H319-H335-H373
Precautionary Statement P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c
EINECSNumber 261-529-2
RTECSNumber OH2915000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only