missing translation for 'onlineSavingsMsg'
Learn More

Beryllium sulfate tetrahydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7049610
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1610414 25 g
AA1610422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1610414 Supplier Thermo Scientific Chemicals Supplier No. 01610414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

In Industries it is used for beryllium salt making, ceramics and chemical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7787-56-6
Molecular Formula BeH8O8S
Molecular Weight (g/mol) 177.128
MDL Number MFCD00149156
InChI Key DIMYTQPLZWDZFE-UHFFFAOYSA-L
Synonym beryllium sulfate tetrahydrate, ccris 85, beryllium sulphate tetrahydrate, beryllium monosulfate tetrahydrate, unii-2tyk3lf8zn, beryllium sulfate, tetrahydrate 1:1:4, 2tyk3lf8zn, sulfuric acid, beryllium salt 1:1 , tetrahydrate, beryllium sulfate tetrahydrate beryllium and beryllium compounds, dsstox_cid_4603
PubChem CID 62672
ChEBI CHEBI:53502
IUPAC Name beryllium;sulfate;tetrahydrate
SMILES [Be+2].O.O.O.O.[O-]S(=O)(=O)[O-]

Specifications

Physical Form Lump
Quantity 25 g
UN Number UN1566
Merck Index 14,1175
Solubility Information Freely soluble in water; Insoluble in alcohol
Formula Weight 177.14 (105.07 Anhydrous)
Percent Purity 99.99%
Density 1.1713 g/mL
Assay (metals basis)
Chemical Name or Material Beryllium sulfate tetrahydrate
Hazard Category H301-H315-H317-H319-H330-H335-H350i-H372
Hazard Statement GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BERYLLIUM COMPOUNDS, N.O.S.
EINECSNumber 236-842-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only