missing translation for 'onlineSavingsMsg'
Learn More

Beryllium, Oil based standard solution, Specpure™ Be 1000μg/g

 

Supplier:  Thermo Scientific Chemicals 01422118

Catalog No. AA1422118


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Beryllium Oil based standard solution
Liquid
50 g
Be 1000μg/g
MFCD00134032
Specpure

Chemical Identifiers

MFCD00134032
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350
May cause cancer.

P201-P202-P281-P301+P310-P308+P313-P331-P501c

H304-H350

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only