missing translation for 'onlineSavingsMsg'
Learn More

Beryllium foil, 0.1mm (0.004in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7051610
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41643FF 25 x 25 mm
AA41643FI 50 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41643FF Supplier Thermo Scientific Chemicals Supplier No. 041643FF
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-41-7
Molecular Formula Be
Molecular Weight (g/mol) 9.01
MDL Number MFCD00134032
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
Synonym glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
PubChem CID 5460467
ChEBI CHEBI:33783
IUPAC Name beryllium
SMILES [Be]

Specifications

Quantity 25 x 25 mm
Odor Odorless
Weight ≈0.12g/25x25mm
Form Foil
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Beryllium foil, 0.1mm (0.004 in.) thick, Vacuum tight
Hazard Category H301-H315-H317-H319-H330-H335-H350i-H372
Hazard Statement GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c
EINECSNumber 231-150-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only