Learn More
Beryllium foil, 0.05mm (0.002in) thick, 99.5% (metals basis), Thermo Scientific Chemicals
$1,320.45 - $1,320.45
Chemical Identifiers
| CAS | 7440-41-7 |
|---|---|
| Molecular Formula | Be |
| Molecular Weight (g/mol) | 9.01 |
| MDL Number | MFCD00134032 |
| InChI Key | ATBAMAFKBVZNFJ-UHFFFAOYSA-N |
| Synonym | glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds |
| PubChem CID | 5460467 |
| ChEBI | CHEBI:33783 |
| IUPAC Name | beryllium |
| SMILES | [Be] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Not vacuum tight
Chemical Identifiers
| 7440-41-7 | |
| 9.01 | |
| ATBAMAFKBVZNFJ-UHFFFAOYSA-N | |
| 5460467 | |
| beryllium |
| Be | |
| MFCD00134032 | |
| glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds | |
| CHEBI:33783 | |
| [Be] |
Specifications
| 7440-41-7 | |
| Odorless | |
| Foil | |
| MFCD00134032 | |
| ATBAMAFKBVZNFJ-UHFFFAOYSA-N | |
| beryllium | |
| 5460467 | |
| 99.5% | |
| (metals basis) |
| 25 x 25 mm | |
| ≈0.058g/25x25mm | |
| Be | |
| glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds | |
| [Be] | |
| 9.01 | |
| CHEBI:33783 | |
| Beryllium foil, 0.05mm (0.002 in.) thick, Not certified pinhole free |
Safety and Handling
GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c
H301-H315-H317-H319-H330-H335-H350i-H372
EINECSNumber : 231-150-7
RTECSNumber : DS1750000
TSCA : Yes
storageNote1 :
- Not vacuum tight
Recommended Storage : Ambient temperatures
RUO – Research Use Only