missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bermekimab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542060
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Bermekimab is under investigation in clinical trial NCT01384630 (Safety, Pharmacokinetics, and Efficacy of RA-18C3 in Subjects With Moderate to Severe Psoriasis).Chemical Identifiers
| Bermekimab Humanized | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human | |
| MABp1; Xilonix | |
| Primary | |
| Protein A |
| 2.34 mg/mL | |
| ELISA, SDS-Page | |
| Unconjugated | |
| Human | |
| RUO | |
| PBS with no preservative; pH 7.5 | |
| Human IL1A. | |
| Antibody |
Specifications
| Bermekimab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MABp1; Xilonix | |
| Human IL1A. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 2.34 mg/mL | |
| PBS with no preservative; pH 7.5 | |
| Human | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |