Learn More
Berkshire™ BCR™ Cleanroom Mopping System: Mop 4
Looped-end construction and fantail design minimize particle generation
Supplier:  Berkshire™ BCM4IR12
 View more versions of this product
Catalog No. 19121756
Only null left
Description
Description
  • Made of chemical- and abrasion-resistant continuous filament knitted polyester
  • Tubular knit construction for cleanliness
  • Laundered and packaged in an ISO Class 4 (formerly Class 10) cleanroom
  • Available gamma irradiated
Specifications
Chemical Identifiers
White
Autoclavable
Easy removal of contamination from floors and for the distribution of disinfectants
Knitted Polyester
20 in.
11.81 in.
ISO Class 4
Specifications
BCR Mop 4 Gamma-irradiated
20 in.
Autoclavable
Easy removal of contamination from floors and for the distribution of disinfectants
Knitted Polyester
White
Sterile
11.81 in.
ISO Class 4
Product Suggestions
Product Suggestions