missing translation for 'onlineSavingsMsg'
Learn More

Bergapten, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014282
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014282 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014282 Supplier Merck Emd Millipore Supplier No. 6966425MG
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 484-20-8
Physical Form Neat
Linear Formula C12H8O4
MDL Number MFCD00010272
Melting Point 189°C to 193°C; 190°C to 193°C
Synonym 5-Methoxypsoralen; 4-Methoxyfuro[3,2-g]benzopyrane-7-one; Bergapten; Geralen; Heraclin; Majudin
Type Analytical Standard
Quantity 25 mg
Formula Weight 216.19
Percent Purity ≥99% (HPLC)
Grade Analytical Standard
Show More Show Less