missing translation for 'onlineSavingsMsg'
Learn More

Berberine Sulfate Hydrate 98.0+%, TCI America™

Catalog No. p-7118453
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04515G 5 g
B045125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B04515G Supplier TCI America Supplier No. B04515G
Only null left

Chemical Identifiers

CAS 316-41-6
Molecular Formula C40H44N2O15S+2
Molecular Weight (g/mol) 824.851
MDL Number MFCD00031743
InChI Key FYBWCLKVKGHJKS-UHFFFAOYSA-N
PubChem CID 129893573
SMILES COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.O.O.O.OS(=O)(=O)O

Specifications

Color Yellow
Quantity 5 g
UN Number 1544
Formula Weight 768.79
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Berberine Sulfate Hydrate
EINECSNumber (9)-1513
RTECSNumber DR9867300
TSCA No