missing translation for 'onlineSavingsMsg'
Learn More

Beraha's Sulfamic Acid Reagent no. 4, Reagents

Catalog No. 01335833
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335833 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335833 Supplier Labchem Inc Supplier No. CB004600500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Beraha's Sulfamic Acid Reagent #4, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1341-49-7 , 7727-21-1 , 7727-21-1
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Color Colorless to Light Yellow
Boiling Point 100°C
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.026
Quantity 500 mL
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Beraha's Sulfamic Acid Reagent 4