missing translation for 'onlineSavingsMsg'
Learn More

Benzylurea, 98%

Catalog No. AAB2494514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2494514 25 g
AAB2494522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2494514 Supplier Thermo Scientific Chemicals Supplier No. B2494514

CAS: 538-32-9 | C8H10N2O | 150.181 g/mol

Benzylurea is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylurea is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 538-32-9
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00007951
InChI Key RJNJWHFSKNJCTB-UHFFFAOYSA-N
Synonym 1-benzylurea, n-benzylurea, benzylcarbamide, urea, phenylmethyl, phenylmethylurea, urea, benzyl, cc-pmlsc-dma-p105, amino-n-benzylamide, benzyl urea, benzyl-urea
PubChem CID 10853
IUPAC Name benzylurea
SMILES C1=CC=C(C=C1)CNC(=O)N

Specifications

Melting Point 146°C to 151°C
Quantity 25 g
Beilstein 2208083
Merck Index 14,1147
Solubility Information Slightly soluble in water.
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material Benzylurea
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 208-689-1
RTECSNumber YS1780000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only