accessibility menu, dialog, popup

UserName

CAS RN 538-32-9

IUPAC Name: benzylurea
Synonyms: benzylurea
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: RJNJWHFSKNJCTB-UHFFFAOYSA-N
SMILES: NC(=O)NCC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)