missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriphenylphosphonium Chloride 98.0+%, TCI America™

Catalog No. B0824500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0824500G 500 g
B082425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0824500G Supplier TCI America Supplier No. B0824500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1100-88-5
Molecular Formula C25H22ClP
Molecular Weight (g/mol) 388.87
MDL Number MFCD00011913
InChI Key USFRYJRPHFMVBZ-UHFFFAOYSA-M
Synonym benzyltriphenylphosphonium chloride, triphenylbenzylphosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride, benzyltriphenylphosphanium chloride, benzyl triphenylphosphonium chloride, benzyl-triphenyl-phosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride 1:1, triphenylbenzylphosphine, chloride, benzyl triphenyl phosphoniumchlorid
PubChem CID 70671
IUPAC Name benzyltriphenylphosphanium chloride
SMILES [Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 500 g
UN Number 3464
Formula Weight 388.87
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltriphenylphosphonium Chloride
EINECSNumber (3)-2608
RTECSNumber TA2416500
TSCA Yes