missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriphenylphosphonium Chloride, 99%

Catalog No. p-3737933
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106210250 25 g Glass bottle
AC106211000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106210250 Supplier Thermo Scientific Chemicals Supplier No. 106210250
Only null left

CAS: 1100-88-5 | C25H22ClP | 388.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 1100-88-5
Molecular Formula C25H22ClP
Molecular Weight (g/mol) 388.87
MDL Number MFCD00011913
InChI Key USFRYJRPHFMVBZ-UHFFFAOYSA-M
Synonym benzyltriphenylphosphonium chloride, triphenylbenzylphosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride, benzyltriphenylphosphanium chloride, benzyl triphenylphosphonium chloride, benzyl-triphenyl-phosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride 1:1, triphenylbenzylphosphine, chloride, benzyl triphenyl phosphoniumchlorid
PubChem CID 70671
IUPAC Name benzyl(triphenyl)phosphanium;chloride
SMILES [Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 337.0°C
Color White
Flash Point >300°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H5CH2P(C6H5)3Cl
Quantity 25 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,slightly soluble in lower aliphatic alcohols
Formula Weight 388.86
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Benzyltriphenylphosphonium chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with wat
EINECSNumber 214-154-3

RUO – Research Use Only