missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriphenylphosphonium chloride, 99%

Catalog No. AAA1403436
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1403436 500 g
AAA1403422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1403436 Supplier Thermo Scientific Chemicals Supplier No. A1403436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1100-88-5 | C25H22ClP | 388.87 g/mol

Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltriphenylphosphonium chloride is used as the accelerator of the Bisphenol AF curing system for fluoroelastomer. Also used as phase-transfer catalysts in the synthesis of certain organic compounds. It is used as a reactant for the synthesis of chemical compounds.

Solubility
Soluble in water and alcohol. Insoluble in acetone.

Notes
Hygroscopic. Store under inert gas. Store away from strong oxidizing agents, water/ moisture..

Chemical Identifiers

CAS 1100-88-5
Molecular Formula C25H22ClP
Molecular Weight (g/mol) 388.87
MDL Number MFCD00011913
InChI Key USFRYJRPHFMVBZ-UHFFFAOYSA-M
Synonym benzyltriphenylphosphonium chloride, triphenylbenzylphosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride, benzyltriphenylphosphanium chloride, benzyl triphenylphosphonium chloride, benzyl-triphenyl-phosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride 1:1, triphenylbenzylphosphine, chloride, benzyl triphenyl phosphoniumchlorid
PubChem CID 70671
IUPAC Name benzyl(triphenyl)phosphanium;chloride
SMILES [Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼330°C (decomposition)
Flash Point >300°C (572°F)
Odor Odorless
Quantity 500 g
UN Number UN3464
Beilstein 3599868
Sensitivity Hygroscopic
Solubility Information Soluble in water and alcohol. Insoluble in acetone.
Formula Weight 388.88
Percent Purity 99%
Chemical Name or Material Benzyltriphenylphosphonium chloride
Show More Show Less
Hazard Category H300+H330-H315-H318-H335
Hazard Statement GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
EINECSNumber 214-154-3
RTECSNumber TA2416500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only