missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium tribromide, 97%, Thermo Scientific™

Chemical Identifiers

CAS 111865-47-5
Molecular Formula C10H16Br3N
Molecular Weight (g/mol) 389.95
MDL Number MFCD00134423
InChI Key GSDDWZVWOXMLJZ-UHFFFAOYSA-N
Synonym benzyltrimethylammonium tribromide
PubChem CID 86222689
IUPAC Name benzyl(trimethyl)azanium;molecular bromine;hydrobromide
SMILES C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC388320100
SDS View Documents
Thermo Scientific Chemicals
388320100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC388320500
SDS View Documents
Thermo Scientific Chemicals
388320500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

111865-47-5
389.95
GSDDWZVWOXMLJZ-UHFFFAOYSA-N
86222689
C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr
C10H16Br3N
MFCD00134423
benzyltrimethylammonium tribromide
benzyl(trimethyl)azanium;molecular bromine;hydrobromide

Specifications

111865-47-5
100.0
96% min. (Iodometry)
C10H16Br3N
10g
Solubility in water: .. Other solubilities: soluble in alcohol
C[N+](C)(C)CC1=CC=CC=C1.Br.BrBr
389.95
389.95
Benzyltrimethylammonium tribromide
96.0
Authentic
Glass bottle
MFCD00134423
benzyltrimethylammonium tribromide
GSDDWZVWOXMLJZ-UHFFFAOYSA-N
benzyl(trimethyl)azanium;molecular bromine;hydrobromide
86222689
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only