accessibility menu, dialog, popup

UserName

CAS RN 111865-47-5

IUPAC Name: benzyltrimethylazanium dibromine hydrobromide
Synonyms: Brom benzyltrimethylazanium hydrobromide
Molecular Weight (g/mol): 390.96
Molecular Formula: C10H17Br3N
InChi Key: GSDDWZVWOXMLJZ-UHFFFAOYSA-N
SMILES: Br.BrBr.C[N+](C)(C)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)