Learn More
Benzyltrimethylammonium hydroxide, 40 wt% solution in methanol
CAS: 100-85-6 | C10H17NO | 167.252 g/mol
Supplier: Thermo Scientific Chemicals 106200010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
| Benzyltrimethylammonium hydroxide | |
| 40 wt. %solution in Methanol | |
| 39.0,59.0 | |
| C10H17NO | |
| MFCD00008281 | |
| 0.9250g/mL | |
| Solubility in water: miscible. | |
| C[N+](C)(C)CC1=CC=CC=C1.[OH-] | |
| 167.252 | |
| 167.25 | |
| Glass bottle | |
| 1 L | |
| Solution |
| Yellow | |
| 100-85-6,67-56-1 | |
| 41.0,61.0 | |
| C6H5CH2N(CH3)3OH | |
| 01,1252; 05,29; 08,36; 12,48; 05,30 | |
| benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide | |
| NDKBVBUGCNGSJJ-UHFFFAOYSA-M | |
| benzyl(trimethyl)azanium;hydroxide | |
| 66854 | |
| ≥40% | |
| 0.925 | |
| 15°C |
Chemical Identifiers
| 100-85-6 | |
| 167.252 | |
| NDKBVBUGCNGSJJ-UHFFFAOYSA-M | |
| 66854 | |
| C[N+](C)(C)CC1=CC=CC=C1.[OH-] |
| C10H17NO | |
| MFCD00008281 | |
| benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide | |
| benzyl(trimethyl)azanium;hydroxide |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes damage to organs.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 202-895-5
RTECSNumber : BO8575000
TSCA : TSCA
RUO – Research Use Only