missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium Hydroxide (10% in Water), TCI America™

Catalog No. B107025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B107025ML 25 mL
B1070500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B107025ML Supplier TCI America Supplier No. B107025ML

Chemical Identifiers

CAS 100-85-6
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00008281
InChI Key NDKBVBUGCNGSJJ-UHFFFAOYSA-M
Synonym benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
PubChem CID 66854
IUPAC Name benzyl(trimethyl)azanium;hydroxide
SMILES C[N+](C)(C)CC1=CC=CC=C1.[OH-]

Specifications

Color Yellow
UN Number 3267
Formula Weight 167.25
Quantity 25 mL
Physical Form Clear Liquid at 20°C
Chemical Name or Material Benzyltrimethylammonium Hydroxide (10% in Water)
EINECSNumber (3)-2694&(9)-0929
RTECSNumber BO8575000
TSCA Yes