missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium Dichloroiodate 97.0+%, TCI America™

Catalog No. B16045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16045G 5 g
B160425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16045G Supplier TCI America Supplier No. B16045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 114971-52-7
Molecular Formula C10H16Cl2IN
Molecular Weight (g/mol) 348.049
MDL Number MFCD00075259
InChI Key PPDJNZTUDFPAHX-UHFFFAOYSA-N
Synonym benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
PubChem CID 2724680
SMILES C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl

Specifications

Melting Point 125°C
Quantity 5 g
Formula Weight 348.05
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltrimethylammonium Dichloroiodate
EINECSNumber (3)-2694
TSCA No