missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium Chloride 99.0+%, TCI America™

Catalog No. p-7118450
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B044725G 25 g
B0447100G 100 g
B0447500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B044725G Supplier TCI America Supplier No. B044725G
Only null left

Chemical Identifiers

CAS 56-93-9
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD00011782
InChI Key KXHPPCXNWTUNSB-UHFFFAOYSA-M
Synonym benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
PubChem CID 5963
IUPAC Name benzyltrimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CC1=CC=CC=C1

Specifications

Melting Point 239°C
Color White
pH 5.0 - 7.0 (50g/L)
Quantity 25 g
UN Number 2811
Formula Weight 185.70
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltrimethylammonium Chloride
EINECSNumber (3)-2694
RTECSNumber BO8400000
TSCA Yes