accessibility menu, dialog, popup

UserName

CAS RN 56-93-9

IUPAC Name: benzyltrimethylazanium chloride
Synonyms: benzyl trimethyl ammonium chloride
Molecular Weight (g/mol): 185.7
Molecular Formula: C10H16ClN
InChi Key: KXHPPCXNWTUNSB-UHFFFAOYSA-M
SMILES: [Cl-].C[N+](C)(C)CC1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (8)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)