missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium chloride, 97%

Catalog No. AAA1675922
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1675922 100 g
AAA1675936 500 g
AAA167590E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1675922 Supplier Thermo Scientific Chemicals Supplier No. A1675922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-93-9 | C10H16ClN | 185.70 g/mol

Benzyltrimethylammonium chloride are Ccommercially important catalyst. Used in antistatic agent, detergent Sanitisers, softner for textiles and paper products, phase transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltrimethylammonium chloride are Ccommercially important catalyst. Used in antistatic agent, detergent Sanitisers, softner for textiles and paper products, phase transfer catalyst.

Solubility
Soluble in water, ethanol, and butanol and slightly soluble in butyl phthalate

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Chemical Identifiers

CAS 56-93-9
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD00011782
InChI Key KXHPPCXNWTUNSB-UHFFFAOYSA-M
Synonym benzyltrimethylammonium chloride, tmbac, benzyl trimethylammonium chloride, benzyltrimethyl ammonium chloride, benzyl trimethyl ammonium chloride, benzenemethanaminium, n,n,n-trimethyl-, chloride, benzyltrimethylazanium chloride, unii-vnk45y7ba1, ammonium, benzyltrimethyl-, chloride, trimethylbenzylammonium chloride
PubChem CID 5963
IUPAC Name benzyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CC1=CC=CC=C1

Specifications

Melting Point ∼238°C (decomposition)
pH 6 to 8
Odor Odorless
Refractive Index 1.444
Quantity 100 g
Beilstein 3917255
Sensitivity Hygroscopic
Solubility Information Soluble in water,ethanol,and butanol and slightly soluble in butyl phthalate
Formula Weight 185.7
Percent Purity 97%
Chemical Name or Material Benzyltrimethylammonium chloride
Show More Show Less
Hazard Category H301+H311-H332-H341-H500
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P312-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 200-300-3
RTECSNumber BO8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only