missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethylammonium Chloride 98.0+%, TCI America™

Catalog No. B044425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B044425G 25 g
B0444500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B044425G Supplier TCI America Supplier No. B044425G

Chemical Identifiers

CAS 56-37-1
Molecular Formula C13H22ClN
Molecular Weight (g/mol) 227.78
MDL Number MFCD00011824
InChI Key HTZCNXWZYVXIMZ-UHFFFAOYSA-M
Synonym benzyltriethylammonium chloride, tebac, benzyl triethylammonium chloride, benzyl triethyl ammonium chloride, benzyltriethylammoniumchloride, triethylbenzylammonium chloride, bteac, teba, benzyltriethylazanium chloride, n-benzyl-n,n-diethylethanaminium chloride
PubChem CID 66133
IUPAC Name benzyltriethylazanium chloride
SMILES [Cl-].CC[N+](CC)(CC)CC1=CC=CC=C1

Specifications

Melting Point 192°C
Color White
pH 4.5 to 7.0 (50g/L H2O soln.)
Quantity 25 g
Formula Weight 227.78
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltriethylammonium Chloride
EINECSNumber (3)-2694
RTECSNumber BO8275000
TSCA Yes