missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethylammonium chloride, 99%

Catalog No. p-7021078
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1326822 100 g
AAA1326836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1326822 Supplier Thermo Scientific Chemicals Supplier No. A1326822

CAS: 56-37-1 | C13H22ClN | 227.78 g/mol

Benzyltriethylammonium chloride is used as a catalyst in the preparation of 2-phenylbutyronitrile from phenyl acetonitrile. It is involved in the Knoevenagel condensation of carbonyl compounds with active methylene compounds to give olefinic products. It acts as a phase transfer catalyst used in the alkylation reaction. It reacts with 1H-Pyridine-2-thione to get 2-Benzylsulfanyl-pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltriethylammonium chloride is used as a catalyst in the preparation of 2-phenylbutyronitrile from phenyl acetonitrile. It is involved in the Knoevenagel condensation of carbonyl compounds with active methylene compounds to give olefinic products. It acts as a phase transfer catalyst used in the alkylation reaction. It reacts with 1H-Pyridine-2-thione to get 2-Benzylsulfanyl-pyridine.

Solubility
Soluble in acetone, alcohol and water. Insoluble in ether.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 56-37-1
Molecular Formula C13H22ClN
Molecular Weight (g/mol) 227.78
MDL Number MFCD00011824
InChI Key HTZCNXWZYVXIMZ-UHFFFAOYSA-M
Synonym benzyltriethylammonium chloride, tebac, benzyl triethylammonium chloride, benzyl triethyl ammonium chloride, benzyltriethylammoniumchloride, triethylbenzylammonium chloride, bteac, teba, benzyltriethylazanium chloride, n-benzyl-n,n-diethylethanaminium chloride
PubChem CID 66133
IUPAC Name benzyl(triethyl)azanium;chloride
SMILES [Cl-].CC[N+](CC)(CC)CC1=CC=CC=C1

Specifications

Melting Point ∼190°C (decomposition)
Quantity 100 g
Beilstein 3574984
Sensitivity Hygroscopic
Solubility Information Soluble in acetone,alcohol and water. Insoluble in ether.
Formula Weight 227.78
Percent Purity 99%
Chemical Name or Material Benzyltriethylammonium chloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 200-270-1
RTECSNumber BO8275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only