missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethylammonium chloride, 98%

Catalog No. p-66112
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164021000 100 g Glass bottle
AC164025000 500 g Glass bottle
AC164020025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164021000 Supplier Thermo Scientific Chemicals Supplier No. 164021000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-37-1 | C13H22ClN | 227.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 56-37-1
Molecular Formula C13H22ClN
Molecular Weight (g/mol) 227.78
MDL Number MFCD00011824
InChI Key HTZCNXWZYVXIMZ-UHFFFAOYSA-M
Synonym benzyltriethylammonium chloride, tebac, benzyl triethylammonium chloride, benzyl triethyl ammonium chloride, benzyltriethylammoniumchloride, triethylbenzylammonium chloride, bteac, teba, benzyltriethylazanium chloride, n-benzyl-n,n-diethylethanaminium chloride
PubChem CID 66133
IUPAC Name benzyl(triethyl)azanium;chloride
SMILES [Cl-].CC[N+](CC)(CC)CC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 185.0°C
Color White to Beige
Flash Point >275°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Packaging Glass bottle
Linear Formula C6H5CH2N(C2H5)3Cl
Quantity 100 g
Solubility Information Solubility in water: 700g/L water (20°C). Other solubilities: soluble in methanol,ethanol,isopropanol,dmf,soluble in acetone,ch2cl2
Formula Weight 227.78
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Benzyltriethylammonium chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-270-1
RTECSNumber BO8275000
TSCA TSCA

RUO – Research Use Only