missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethoxysilane 96.0+%, TCI America™

Catalog No. B32825G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32825G 5 g
B32821G 1 g
B328225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B32825G Supplier TCI America Supplier No. B32825G
Only null left

Chemical Identifiers

CAS 2549-99-7
Molecular Formula C13H22O3Si
Molecular Weight (g/mol) 254.40
MDL Number MFCD00026751
InChI Key CPLASELWOOUNGW-UHFFFAOYSA-N
PubChem CID 75690
IUPAC Name benzyltriethoxysilane
SMILES CCO[Si](CC1=CC=CC=C1)(OCC)OCC

Specifications

Color Yellow
Boiling Point 248°C
Quantity 5 g
Formula Weight 254.40
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyltriethoxysilane