accessibility menu, dialog, popup

UserName

CAS RN 2549-99-7

H3COSiOH3COCH3

CAS RN 2549-99-7

IUPAC Name: benzyltriethoxysilane
Synonyms: benzyltriethoxysilane
Molecular Weight (g/mol): 254.4
Molecular Formula: C13H22O3Si
InChi Key: CPLASELWOOUNGW-UHFFFAOYSA-N
SMILES: CCO[Si](CC1=CC=CC=C1)(OCC)OCC
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results