missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltriethoxysilane 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 248°C |
| Quantity | 1 g |
| Formula Weight | 254.40 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Benzyltriethoxysilane |
Safety and Handling
| TSCA | Yes |