missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltributylammonium Chloride 98.0+%, TCI America™
$29.83 - $182.68
Chemical Identifiers
| CAS | 23616-79-7 |
|---|---|
| Molecular Formula | C19H34ClN |
| Molecular Weight (g/mol) | 311.94 |
| MDL Number | MFCD00011849 |
| InChI Key | VJGNLOIQCWLBJR-UHFFFAOYSA-M |
| Synonym | benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac |
| PubChem CID | 159952 |
| IUPAC Name | benzyltributylazanium chloride |
| SMILES | [Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1 |
Chemical Identifiers
| 23616-79-7 | |
| 311.94 | |
| VJGNLOIQCWLBJR-UHFFFAOYSA-M | |
| 159952 | |
| [Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1 |
| C19H34ClN | |
| MFCD00011849 | |
| benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac | |
| benzyltributylazanium chloride |
Specifications
| 23616-79-7 | |
| White | |
| MFCD00011849 | |
| 1759 | |
| VJGNLOIQCWLBJR-UHFFFAOYSA-M | |
| benzyltributylazanium chloride | |
| 159952 | |
| ≥98.0% (T) | |
| Benzyltributylammonium Chloride |
| 164°C | |
| C19H34ClN | |
| 25 g | |
| benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac | |
| [Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1 | |
| 311.94 | |
| 311.94 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2694
TSCA : Yes