accessibility menu, dialog, popup

UserName

ClCH3NH3CCH3

CAS RN 23616-79-7

IUPAC Name: benzyltributylazanium chloride
Synonyms: benzyltributylazanium chloride
Molecular Weight (g/mol): 311.94
Molecular Formula: C19H34ClN
InChi Key: VJGNLOIQCWLBJR-UHFFFAOYSA-M
SMILES: [Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)