missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltributylammonium Chloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 164°C |
| Color | White |
| Quantity | 25 g |
| UN Number | 1759 |
| Formula Weight | 311.94 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Benzyltributylammonium Chloride |
Safety and Handling
| EINECSNumber | (3)-2694 |
| TSCA | Yes |