missing translation for 'onlineSavingsMsg'
Learn More

Benzyltributylammonium chloride, 98+%

Catalog No. p-66081
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186151000 100 g Glass bottle
AC186155000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC186151000 Supplier Thermo Scientific Chemicals Supplier No. 186151000
Only null left

CAS: 23616-79-7 | C19H34ClN | 311.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 23616-79-7
Molecular Formula C19H34ClN
Molecular Weight (g/mol) 311.94
MDL Number MFCD00011849
InChI Key VJGNLOIQCWLBJR-UHFFFAOYSA-M
Synonym benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac
PubChem CID 159952
IUPAC Name benzyl(tributyl)azanium;chloride
SMILES [Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 155°C to 163°C
Color White to Ivory
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H5CH2N[(CH2)3CH3]3Cl
Quantity 100 g
Fieser 06,41
Solubility Information Solubility in water: freely soluble in water
Formula Weight 311.94
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Benzyltributylammonium chloride, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 245-787-3
TSCA TSCA

RUO – Research Use Only