missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltributylammonium Bromide 98.0+%, TCI America™
$70.84 - $615.14
Chemical Identifiers
| CAS | 25316-59-0 |
|---|---|
| Molecular Formula | C19H34BrN |
| Molecular Weight (g/mol) | 356.392 |
| MDL Number | MFCD00011848 |
| InChI Key | UDYGXWPMSJPFDG-UHFFFAOYSA-M |
| Synonym | benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801 |
| PubChem CID | 2724282 |
| IUPAC Name | benzyl(tributyl)azanium;bromide |
| SMILES | CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-] |
Chemical Identifiers
| 25316-59-0 | |
| 356.392 | |
| UDYGXWPMSJPFDG-UHFFFAOYSA-M | |
| 2724282 | |
| CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-] |
| C19H34BrN | |
| MFCD00011848 | |
| benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801 | |
| benzyl(tributyl)azanium;bromide |
Specifications
| 25316-59-0 | |
| C19H34BrN | |
| 25 g | |
| UDYGXWPMSJPFDG-UHFFFAOYSA-M | |
| benzyl(tributyl)azanium;bromide | |
| 2724282 | |
| ≥98.0% (T) | |
| Benzyltributylammonium Bromide |
| White | |
| MFCD00011848 | |
| benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801 | |
| CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-] | |
| 356.392 | |
| 356.39 | |
| Crystalline Powder |
Safety and Handling
TSCA : No