missing translation for 'onlineSavingsMsg'
Learn More

Benzyltributylammonium Bromide 98.0+%, TCI America™

Catalog No. B1217500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1217500G 500 g
B121725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1217500G Supplier TCI America Supplier No. B1217500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25316-59-0
Molecular Formula C19H34BrN
Molecular Weight (g/mol) 356.392
MDL Number MFCD00011848
InChI Key UDYGXWPMSJPFDG-UHFFFAOYSA-M
Synonym benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801
PubChem CID 2724282
IUPAC Name benzyl(tributyl)azanium;bromide
SMILES CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]

Specifications

Color White
Quantity 500 g
Formula Weight 356.39
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltributylammonium Bromide