missing translation for 'onlineSavingsMsg'
Learn More

Benzyltri-n-butylammonium chloride, 98%

CAS: 23616-79-7 | C19H34ClN | 311.94 g/mol

$213.60 - $711.42

Chemical Identifiers

CAS 23616-79-7
Molecular Formula C19H34ClN
Molecular Weight (g/mol) 311.94
MDL Number MFCD00011849
InChI Key VJGNLOIQCWLBJR-UHFFFAOYSA-M
Synonym benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac
PubChem CID 159952
IUPAC Name benzyl(tributyl)azanium;chloride
SMILES [Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1034536
View Documents
Thermo Scientific Chemicals
A1034536
500 g
Each for $213.60
Only null left
 
AAA103450E
View Documents
Thermo Scientific Chemicals
A103450E
2500 g
Each for $711.42
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23616-79-7
311.94
VJGNLOIQCWLBJR-UHFFFAOYSA-M
159952
[Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1
C19H34ClN
MFCD00011849
benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac
benzyl(tributyl)azanium;chloride

Specifications

23616-79-7
C19H34ClN
500 g
Hygroscopic
VJGNLOIQCWLBJR-UHFFFAOYSA-M
benzyl(tributyl)azanium;chloride
159952
98%
156°C to 162°C
MFCD00011849
3776210
benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac
[Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1
311.94
311.94
Benzyltri-n-butylammonium chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H500

EINECSNumber : 245-787-3

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only