missing translation for 'onlineSavingsMsg'
Learn More

Benzyltri-n-butylammonium chloride, 98%

Catalog No. AAA1034536
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1034536 500 g
AAA103450E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1034536 Supplier Thermo Scientific Chemicals Supplier No. A1034536
Only null left

CAS: 23616-79-7 | C19H34ClN | 311.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23616-79-7
Molecular Formula C19H34ClN
Molecular Weight (g/mol) 311.94
MDL Number MFCD00011849
InChI Key VJGNLOIQCWLBJR-UHFFFAOYSA-M
Synonym benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac
PubChem CID 159952
IUPAC Name benzyl(tributyl)azanium;chloride
SMILES [Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1

Specifications

Melting Point 156°C to 162°C
Quantity 500 g
Beilstein 3776210
Sensitivity Hygroscopic
Formula Weight 311.94
Percent Purity 98%
Chemical Name or Material Benzyltri-n-butylammonium chloride
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 245-787-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only