missing translation for 'onlineSavingsMsg'
Learn More

Benzyltri-n-butylammonium bromide, 99%

CAS: 25316-59-0 | C19H34BrN | 356.392 g/mol

$80.15 - $407.78

Chemical Identifiers

CAS 25316-59-0
Molecular Formula C19H34BrN
Molecular Weight (g/mol) 356.392
MDL Number MFCD00011848
InChI Key UDYGXWPMSJPFDG-UHFFFAOYSA-M
Synonym benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801
PubChem CID 2724282
IUPAC Name benzyl(tributyl)azanium;bromide
SMILES CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2394114
View Documents
Thermo Scientific Chemicals
B2394114
25 g N/A
Only null left
 
AAB2394118
View Documents
Thermo Scientific Chemicals
B2394118
50 g N/A
Only null left
 
AAB2394130
View Documents
Thermo Scientific Chemicals
B2394130
250 g N/A
Only null left
 
Description

Description

Benzyltri-n-butylammonium bromide will react with N-vinyl-phthalimide to produce N-trans-cinnamyl-phthalimide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltri-n-butylammonium bromide will react with N-vinyl-phthalimide to produce N-trans-cinnamyl-phthalimide.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature.
Specifications

Chemical Identifiers

25316-59-0
356.392
UDYGXWPMSJPFDG-UHFFFAOYSA-M
2724282
CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]
C19H34BrN
MFCD00011848
benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801
benzyl(tributyl)azanium;bromide

Specifications

25316-59-0
C19H34BrN
25 g
Hygroscopic
Soluble in water.
CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]
356.392
356.4
Benzyltri-n-butylammonium bromide
∼175°C (decomposition)
MFCD00011848
3776294
benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801
UDYGXWPMSJPFDG-UHFFFAOYSA-M
benzyl(tributyl)azanium;bromide
2724282
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 246-819-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only