missing translation for 'onlineSavingsMsg'
Learn More

Benzylthiourea 98.0+%, TCI America™

Catalog No. B46125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46125G 5 g
B46121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46125G Supplier TCI America Supplier No. B46125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 621-83-0
Molecular Formula C8H10N2S
Molecular Weight (g/mol) 166.24
MDL Number MFCD00041370
InChI Key UCGFRIAOVLXVKL-UHFFFAOYSA-N
Synonym 1-benzylthiourea, 1-benzyl-2-thiourea, n-benzylthiourea, thiourea, phenylmethyl, urea, 1-benzyl-2-thio, unii-mrp6y7412k, amino benzylamino methane-1-thione, n-benzylthioformamide, benzylthioharnstoff, n-benzyl thiourea
PubChem CID 737375
IUPAC Name benzylthiourea
SMILES NC(=S)NCC1=CC=CC=C1

Specifications

Melting Point 164°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 166.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Benzylthiourea